Assay Details
Target Gene Details
Entrez Gene ID: | 140462 |
Gene Name: | ankyrin repeat and SOCS box containing 9 |
Gene Aliases: |
- |
Location: |
Chr.X:15243987-15270476 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB9 | NM_001031739.2 | 7 | 1548 | NP_001026909.1 |
| NM_001168530.1 | 7 | 1579 | NP_001162002.1 | |
| NM_001168531.1 | 8 | 1796 | NP_001162003.1 | |
| NM_024087.2 | 7 | 1609 | NP_076992.1 | |
| XM_005274446.1 | 8 | 1735 | XP_005274503.1 | |
| XM_011545458.1 | 8 | 1648 | XP_011543760.1 | |
| XM_011545460.1 | 8 | 1709 | XP_011543762.1 | |
| XM_017029283.1 | 7 | 1515 | XP_016884772.1 | |
| XM_017029284.1 | 7 | 1428 | XP_016884773.1 | |
| XM_017029285.1 | 7 | 1576 | XP_016884774.1 | |
| AK000643.1 | 7 | 1499 | BAA91302.1 | |
| AK292270.1 | ||||
| AL080091.2 | 7 | 1325 | CAB45706.2 | |
| BC001244.1 | 7 | 1459 | AAH01244.1 | |
| BC013172.1 | 7 | 1498 | AAH13172.1 |
More Information
Set Membership: |
|

Genomic Map