Assay Details
Target Gene Details
Entrez Gene ID: | 79661 |
Gene Name: | nei like DNA glycosylase 1 |
Gene Aliases: |
FPG1, NEI1, hFPG1 |
Location: |
Chr.15:75346895-75355251 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEIL1 | NM_001256552.1 | 2 | 759 | NP_001243481.1 |
| NM_024608.3 | 2 | 835 | NP_078884.2 | |
| NR_046311.1 | 3 | 958 | ||
| XM_005254659.4 | 2 | 695 | XP_005254716.1 | |
| XM_006720677.3 | 2 | 680 | XP_006720740.1 | |
| XM_006720678.3 | 2 | 710 | XP_006720741.1 | |
| XM_006720679.3 | 2 | 618 | XP_006720742.1 | |
| XM_006720680.1 | 3 | 850 | XP_006720743.1 | |
| XM_006720681.1 | 3 | 697 | XP_006720744.1 | |
| XM_011522001.2 | 2 | 600 | XP_011520303.2 | |
| XM_011522002.1 | 3 | 823 | XP_011520304.1 | |
| XM_011522003.2 | 4 | 879 | XP_011520305.1 | |
| XM_011522004.2 | 2 | 567 | XP_011520306.1 | |
| XM_011522005.1 | 2 | 754 | XP_011520307.1 | |
| XM_011522006.1 | 2 | 754 | XP_011520308.1 | |
| XM_011522007.1 | 3 | 850 | XP_011520309.1 | |
| XM_011522008.1 | 3 | 823 | XP_011520310.1 | |
| XM_017022566.1 | 2 | 682 | XP_016878055.1 | |
| XM_017022567.1 | 2 | 514 | XP_016878056.1 | |
| XM_017022568.1 | 2 | 717 | XP_016878057.1 | |
| AB079068.1 | 1 | 329 | BAC06476.1 | |
| AK026055.1 | 2 | 756 | BAB15337.1 | |
| AK097008.1 | 3 | 958 | ||
| AK128372.1 | 3 | 430 | ||
| BC010876.1 | 2 | 740 | AAH10876.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2621n100 | Chr.15:75101872 - 75382048 on Build GRCh38 | Gain |
|
| esv2749894 | Chr.15:75326873 - 75749895 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map