Assay Details
Target Gene Details
Entrez Gene ID: | 350 |
Gene Name: | apolipoprotein H |
Gene Aliases: |
B2G1, B2GP1, BG |
Location: |
Chr.17:66212029-66229438 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOH | NM_000042.2 | NP_000033.2 | ||
| AK298416.1 | ||||
| AK313838.1 | ||||
| BC020703.1 | AAH20703.1 | |||
| BC026283.1 | AAH26283.1 | |||
| BG566874.1 | ||||
| M62839.1 | AAA51766.1 | |||
| S80305.1 | AAB21330.1 | |||
| X53595.1 | CAA37664.1 | |||
| X57847.1 | CAA40977.1 | |||
| X58100.1 | CAA41113.1 |
More Information
Set Membership: |
|

Genomic Map