Assay Details
Target Gene Details
Entrez Gene ID: | 6175 |
Gene Name: | ribosomal protein lateral stalk subunit P0 |
Gene Aliases: |
L10E, LP0, P0, PRLP0, RPP0 |
Location: |
Chr.12:120196700-120201211 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPLP0 | NM_001002.3 | NP_000993.1 | ||
| NM_053275.3 | NP_444505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv976653 | Chr.12:120200045 - 120201198 on Build GRCh38 | Duplication |
|
| nsv832530 | Chr.12:120148265 - 120320372 on Build GRCh38 | Loss |
|
| nsv519657 | Chr.12:120095892 - 120238287 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map