Assay Details
Target Gene Details
Entrez Gene ID: | 55520 |
Gene Name: | elaC ribonuclease Z 1 |
Gene Aliases: |
D29 |
Location: |
Chr.18:50968017-50988120 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELAC1 | NM_018696.2 | 4 | 1522 | NP_061166.1 |
| AB029151.1 | 4 | 1462 | BAA96799.1 | |
| AF308695.1 | 4 | 1506 | AAG24917.1 |
More Information
Set Membership: |
|

Genomic Map