Assay Details
Target Gene Details
Entrez Gene ID: | 102724050 |
Gene Name: | uncharacterized LOC102724050 |
Gene Aliases: |
- |
Location: |
Chr.12:54353691-54467030 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724050 | NR_120486.1 | |||
| NR_120487.1 | ||||
| DA746386.1 | ||||
| DA746474.1 | ||||
| DA757660.1 |
Target Gene Details
Entrez Gene ID: | 25946 |
Gene Name: | zinc finger protein 385A |
Gene Aliases: |
HZF, RZF, ZFP385, ZNF385 |
Location: |
Chr.12:54369129-54391609 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385A | NM_001130967.2 | 8 | 1127 | NP_001124439.1 |
| NM_001130968.2 | 7 | 884 | NP_001124440.1 | |
| NM_001290001.1 | 6 | 979 | NP_001276930.1 | |
| NM_001290002.1 | 9 | 1317 | NP_001276931.1 | |
| NM_001290004.1 | 9 | 1272 | NP_001276933.1 | |
| NM_015481.2 | 7 | 1222 | NP_056296.1 | |
| XM_005268783.4 | 8 | 1149 | XP_005268840.1 | |
| XM_006719342.2 | 9 | 1320 | XP_006719405.1 | |
| XM_011538168.2 | 9 | 1239 | XP_011536470.1 | |
| XM_011538169.2 | 8 | 1143 | XP_011536471.1 | |
| XM_011538170.1 | 7 | 992 | XP_011536472.1 | |
| XM_011538171.2 | 8 | 1287 | XP_011536473.1 | |
| XM_017019174.1 | 8 | 1455 | XP_016874663.1 | |
| AB593085.1 | 9 | 1182 | ||
| AF304052.1 | 7 | 1189 | AAL08625.1 | |
| AK024404.1 | 7 | 860 | BAB14910.1 | |
| AK092724.1 | 8 | 996 | ||
| AK296564.1 | 6 | 922 | ||
| AK315613.1 | 8 | 1120 | ||
| AL117462.1 | 6 | 832 | CAB55938.1 | |
| AY461717.1 | 9 | 1297 | AAS19275.1 | |
| BC029752.1 | 8 | 1108 | AAH29752.1 | |
| CR457327.1 | 6 | 823 | CAG33608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559000 | Chr.12:54343917 - 54396183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map