Assay Details
Target Gene Details
Entrez Gene ID: | 285315 |
Gene Name: | chromosome 3 open reading frame 33 |
Gene Aliases: |
AC3-33 |
Location: |
Chr.3:155762612-155806287 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf33 | NM_001308229.1 | 3 | 269 | NP_001295158.1 |
| NM_173657.2 | 4 | 396 | NP_775928.1 | |
| XM_005247379.4 | 3 | 1241 | XP_005247436.1 | |
| XM_011512710.2 | 2 | 194 | XP_011511012.1 | |
| XM_011512711.2 | 3 | 373 | XP_011511013.1 | |
| AF115515.1 | 3 | 232 | AAO06952.1 | |
| AK055701.1 | 4 | 375 | BAB70989.1 | |
| AK289890.1 | 3 | 269 | ||
| AL832165.1 | 3 | 254 | ||
| BC042198.1 | 3 | 231 | AAH42198.1 | |
| BC064898.1 | 3 | 241 | AAH64898.1 | |
| DA275323.1 | 3 | 269 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3598285 | Chr.3:155760377 - 155780857 on Build GRCh38 | Gain |
|
| dgv4928n100 | Chr.3:155763308 - 155816827 on Build GRCh38 | Gain |
|
| esv25006 | Chr.3:155761458 - 155794985 on Build GRCh38 | Gain |
|
| esv3598283 | Chr.3:155759067 - 155869415 on Build GRCh38 | Loss |
|
| nsv592079 | Chr.3:155757090 - 155776126 on Build GRCh38 | Gain |
|
| nsv516712 | Chr.3:155763820 - 155776126 on Build GRCh38 | Gain |
|
| esv3575620 | Chr.3:155757648 - 155796064 on Build GRCh38 | Gain |
|
| dgv4926n100 | Chr.3:155757649 - 155797695 on Build GRCh38 | Gain |
|
| nsv592080 | Chr.3:155761433 - 155794055 on Build GRCh38 | Gain |
|
| esv33346 | Chr.3:155763431 - 155780652 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map