Assay Details
Target Gene Details
Entrez Gene ID: | 4037 |
Gene Name: | LDL receptor related protein 3 |
Gene Aliases: |
- |
Location: |
Chr.19:33194693-33208867 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRP3 | NM_002333.3 | 7 | 2175 | NP_002324.2 |
| XM_005258945.1 | 7 | 2137 | XP_005259002.1 | |
| AB009462.1 | 7 | 2308 | BAA32330.1 | |
| AF085860.1 | 1 | 216 | ||
| AK074751.1 | 5 | 1891 | ||
| AK126750.1 | 3 | 2983 | ||
| AK129963.1 | 4 | 2579 | BAC85262.1 | |
| AK296536.1 | 7 | 2056 | ||
| AK316314.1 | 6 | 2002 | ||
| BC007408.2 | 7 | 2104 | AAH07408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833808 | Chr.19:33130887 - 33350702 on Build GRCh38 | Loss |
|
| nsv953277 | Chr.19:33192995 - 33270394 on Build GRCh38 | Deletion |
|
| nsv1077340 | Chr.19:33200493 - 33208094 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map