Assay Details
Target Gene Details
Entrez Gene ID: | 84193 |
Gene Name: | SET domain containing 3 |
Gene Aliases: |
C14orf154 |
Location: |
Chr.14:99397746-99486458 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD3 | NM_032233.2 | 13 | 2804 | NP_115609.2 |
| XM_005268127.3 | 12 | 2657 | XP_005268184.1 | |
| XM_011537231.2 | 13 | 2756 | XP_011535533.2 | |
| XM_011537232.2 | 13 | 3934 | XP_011535534.1 | |
| XM_011537233.2 | 11 | 2582 | XP_011535535.1 | |
| XM_011537234.1 | 12 | 2524 | XP_011535536.1 | |
| XM_017021699.1 | 12 | 2652 | XP_016877188.1 | |
| XM_017021700.1 | 13 | 2840 | XP_016877189.1 | |
| XM_017021701.1 | 11 | 2632 | XP_016877190.1 | |
| XM_017021702.1 | 10 | 2451 | XP_016877191.1 | |
| AL359581.2 | 8 | 1994 | CAH56365.1 | |
| BC142995.1 | 13 | 2816 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832871 | Chr.14:99215657 - 99443618 on Build GRCh38 | Gain |
|
| nsv832872 | Chr.14:99216597 - 99417613 on Build GRCh38 | Loss |
|
| esv3635481 | Chr.14:99277609 - 99600560 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map