Assay Details
Target Gene Details
Entrez Gene ID: | 80306 |
Gene Name: | mediator complex subunit 28 |
Gene Aliases: |
1500003D12Rik, EG1, magicin |
Location: |
Chr.4:17614628-17625628 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED28 | NM_025205.4 | 4 | 851 | NP_079481.2 |
| AF317678.1 | ||||
| AF317679.1 | 4 | 828 | ||
| AF318059.1 | 4 | 848 | AAL26906.1 | |
| AF358829.1 | 4 | 836 | AAK32724.1 | |
| BC011936.2 | 4 | 828 | AAH11936.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76919299] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map