Assay Details
Target Gene Details
Entrez Gene ID: | 197021 |
Gene Name: | lactase like |
Gene Aliases: |
KLG, KLPH |
Location: |
Chr.15:66547468-66566025 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCTL | NM_001278562.1 | 8 | 961 | NP_001265491.1 |
| NM_207338.3 | 9 | 1252 | NP_997221.2 | |
| NR_103739.1 | 7 | 974 | ||
| XM_017021999.1 | 9 | 1171 | XP_016877488.1 | |
| AK090598.1 | 8 | 946 | ||
| AK311441.1 | 7 | 974 | ||
| AY358729.1 | 9 | 1252 | AAQ89091.1 |
More Information
Set Membership: |
|

Genomic Map