Assay Details
Target Gene Details
Entrez Gene ID: | 55816 |
Gene Name: | docking protein 5 |
Gene Aliases: |
C20orf180, IRS-6, IRS6 |
Location: |
Chr.20:54475472-54651171 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOK5 | NM_001294161.1 | 1 | 381 | NP_001281090.1 |
| NM_018431.4 | 1 | 381 | NP_060901.2 | |
| NM_177959.2 | 1 | 135 | NP_808874.1 | |
| AF132732.1 | 1 | 103 | AAF66443.1 | |
| AI822030.1 | ||||
| AK313222.1 | 1 | 93 | ||
| BC008992.1 | 1 | 135 | AAH08992.1 | |
| BI758267.1 | 1 | 126 | ||
| DA161365.1 | 1 | 381 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586224 | Chr.20:53858311 - 54662951 on Build GRCh38 | Gain |
|
| dgv67n68 | Chr.20:54241802 - 54480638 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map