Assay Details
Target Gene Details
Entrez Gene ID: | 810 |
Gene Name: | calmodulin like 3 |
Gene Aliases: |
CLP |
Location: |
Chr.10:5524889-5526268 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALML3 | NM_005185.3 | 1 | 925 | NP_005176.1 |
| BC031889.1 | 1 | 833 | AAH31889.1 | |
| CU454493.1 | 1 | 273 | ||
| M36707.1 | 1 | 1805 | ||
| M58026.1 | 1 | 854 | AAA36356.1 |
Target Gene Details
Entrez Gene ID: | 100132159 |
Gene Name: | CALML3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:5514244-5526246 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALML3-AS1 | NR_120496.1 | |||
| NR_120497.1 | ||||
| DA220455.1 | ||||
| DA770774.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831774 | Chr.10:5421593 - 5615458 on Build GRCh38 | Gain |
|
| nsv549886 | Chr.10:5163474 - 5590097 on Build GRCh38 | Loss |
|
| nsv523491 | Chr.10:5500146 - 5678104 on Build GRCh38 | Gain |
|
| nsv1051283 | Chr.10:5463511 - 5577978 on Build GRCh38 | Gain |
|
| esv2759729 | Chr.10:5515061 - 5762817 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map