Assay Details
Target Gene Details
Entrez Gene ID: | 56890 |
Gene Name: | Mdm1 nuclear protein |
Gene Aliases: |
- |
Location: |
Chr.12:68294566-68332381 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDM1 | NM_001205028.1 | 8 | 1147 | NP_001191957.1 |
| NM_017440.4 | 8 | 1252 | NP_059136.2 | |
| XM_005269026.1 | 9 | 1207 | XP_005269083.1 | |
| XM_011538569.1 | 9 | 1207 | XP_011536871.1 | |
| XM_017019646.1 | 8 | 2553 | XP_016875135.1 | |
| XM_017019647.1 | 10 | 1283 | XP_016875136.1 | |
| XM_017019648.1 | 9 | 1714 | XP_016875137.1 | |
| XM_017019649.1 | 9 | 1144 | XP_016875138.1 | |
| XM_017019650.1 | 8 | 3064 | XP_016875139.1 | |
| AK026745.1 | 3 | 309 | ||
| AK297311.1 | 8 | 1052 | ||
| BC028355.2 | 8 | 1203 | AAH28355.1 |
More Information
Set Membership: |
|

Genomic Map