Assay Details
Target Gene Details
Entrez Gene ID: | 1408 |
Gene Name: | cryptochrome circadian clock 2 |
Gene Aliases: |
HCRY2, PHLL2 |
Location: |
Chr.11:45847118-45883248 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRY2 | NM_001127457.2 | 8 | 1227 | NP_001120929.1 |
| NM_021117.3 | 8 | 1363 | NP_066940.2 | |
| AB014558.1 | 8 | 1267 | BAA31633.1 | |
| AK291260.1 | 8 | 1294 | ||
| AK294904.1 | 8 | 1221 | ||
| AK295627.1 | 4 | 763 | ||
| AK302865.1 | 8 | 1363 | ||
| BC035161.1 | 10 | 1626 | AAH35161.1 | |
| BC041814.1 | 8 | 1291 | AAH41814.1 | |
| EU219622.1 | 8 | 1286 |
More Information
Set Membership: |
|

Genomic Map