Assay Details
Target Gene Details
Entrez Gene ID: | 563 |
Gene Name: | alpha-2-glycoprotein 1, zinc-binding |
Gene Aliases: |
ZA2G, ZAG |
Location: |
Chr.7:99966727-99976112 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AZGP1 | NM_001185.3 | NP_001176.1 | ||
| AA583051.1 | ||||
| AI678968.1 | ||||
| AW015837.1 | ||||
| BC005306.1 | AAH05306.1 | |||
| BC033830.1 | AAH33830.1 | |||
| CB305376.1 | ||||
| D90427.1 | BAA14417.1 | |||
| M76707.1 | AAA61311.1 | |||
| X59766.1 | CAA42438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv433391 | Chr.7:99966338 - 100020983 on Build GRCh38 | Gain |
|
| nsv824234 | Chr.7:99956111 - 99987801 on Build GRCh38 | Loss |
|
| esv23518 | Chr.7:99966510 - 100027788 on Build GRCh38 | Gain |
|
| nsv824233 | Chr.7:99955948 - 99973377 on Build GRCh38 | Loss |
|
| nsv607930 | Chr.7:99927618 - 100004568 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map