Assay Details
Target Gene Details
Entrez Gene ID: | 11166 |
Gene Name: | SRY-box 21 |
Gene Aliases: |
SOX25 |
Location: |
Chr.13:94709622-94712543 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX21 | NM_007084.3 | 1 | 709 | NP_009015.1 |
| AI569726.1 | 1 | 159 | ||
| AI816765.1 | 1 | 184 | ||
| CX869660.1 | 1 | 193 | ||
| DB493621.1 | 1 | 260 | ||
| X65666.1 | CAA46617.1 | |||
| X71136.1 | 1 | 182 | CAA50466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070605 | Chr.13:94711245 - 94714046 on Build GRCh38 | Deletion |
|
| nsv832686 | Chr.13:94554488 - 94748738 on Build GRCh38 | Gain |
|
| esv28079 | Chr.13:94710850 - 94713584 on Build GRCh38 | Loss |
|
| esv34004 | Chr.13:94254283 - 94740390 on Build GRCh38 | Loss |
|
| nsv826750 | Chr.13:94706248 - 94715816 on Build GRCh38 | Gain |
|
| dgv3325n54 | Chr.13:94711363 - 94713127 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map