Assay Details
Target Gene Details
Entrez Gene ID: | 160428 |
Gene Name: | aldehyde dehydrogenase 1 family member L2 |
Gene Aliases: |
mtFDH |
Location: |
Chr.12:105019784-105084609 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1L2 | NM_001034173.3 | NP_001029345.2 | ||
| NR_027752.1 | ||||
| XM_011537986.2 | XP_011536288.1 | |||
| XM_017018889.1 | XP_016874378.1 | |||
| AF086109.1 | ||||
| AK300373.1 | ||||
| BC103934.1 | AAI03935.1 | |||
| BC103935.1 | AAI03936.1 | |||
| CR627287.1 | CAH10368.1 | |||
| CR749561.1 | CAH18358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832505 | Chr.12:104921574 - 105103987 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map