Assay Details
Target Gene Details
Entrez Gene ID: | 138199 |
Gene Name: | carnosine N-methyltransferase 1 |
Gene Aliases: |
C9orf41, UPF0586 |
Location: |
Chr.9:74980794-75028456 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARNMT1 | NM_001320497.1 | 3 | 349 | NP_001307426.1 |
| NM_152420.2 | 3 | 669 | NP_689633.1 | |
| NR_135282.1 | 3 | 669 | ||
| XM_005251701.3 | 3 | 344 | XP_005251758.2 | |
| XM_011518223.2 | 3 | 695 | XP_011516525.1 | |
| XM_017014269.1 | 3 | 656 | XP_016869758.1 | |
| XM_017014270.1 | 4 | 602 | XP_016869759.1 | |
| XM_017014271.1 | 4 | 1094 | XP_016869760.1 | |
| AK098661.1 | 2 | 96 | BAC05369.1 | |
| AK299691.1 | 3 | 512 | ||
| BC034033.1 | 3 | 633 | AAH34033.1 | |
| BI560441.1 | 3 | 673 | ||
| BX538061.1 | 2 | 186 | CAD97992.1 | |
| BX647386.1 | 3 | 438 |
More Information
Set Membership: |
|

Genomic Map