Assay Details
Target Gene Details
Entrez Gene ID: | 2167 |
Gene Name: | fatty acid binding protein 4 |
Gene Aliases: |
A-FABP, AFABP, ALBP, HEL-S-104, aP2 |
Location: |
Chr.8:81478497-81483238 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FABP4 | NM_001442.2 | NP_001433.1 | ||
| BC003672.1 | AAH03672.1 | |||
| BQ880795.1 | ||||
| BT006809.1 | AAP35455.1 | |||
| CD000452.1 | ||||
| CR456903.1 | CAG33184.1 | |||
| FJ224319.1 | ||||
| J02874.1 | AAA51689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv465721 | Chr.8:81405621 - 81768032 on Build GRCh38 | Gain |
|
| nsv611608 | Chr.8:81464464 - 81480614 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112313579] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map