Assay Details
Target Gene Details
Entrez Gene ID: | 9588 |
Gene Name: | peroxiredoxin 6 |
Gene Aliases: |
1-Cys, AOP2, HEL-S-128m, NSGPx, PRX, aiPLA2, p29 |
Location: |
Chr.1:173477347-173488807 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDX6 | NM_004905.2 | NP_004896.1 | ||
| AJ844621.1 | CAH59743.1 | |||
| AK289352.1 | ||||
| AK300680.1 | ||||
| BC035857.1 | AAH35857.1 | |||
| BC053550.1 | AAH53550.1 | |||
| D14662.1 | BAA03496.1 | |||
| EU794652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831915 | Chr.1:173299285 - 173513018 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map