Assay Details
Target Gene Details
Entrez Gene ID: | 162417 |
Gene Name: | N-acetylglutamate synthase |
Gene Aliases: |
AGAS, ARGA |
Location: |
Chr.17:44004546-44009068 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAGS | NM_153006.2 | NP_694551.1 | ||
| XM_011524438.1 | XP_011522740.1 | |||
| XM_011524439.2 | XP_011522741.1 | |||
| AK074407.1 | ||||
| AK091938.1 | 2 | 375 | ||
| AK300923.1 | ||||
| AK314432.1 | ||||
| AY116538.1 | AAM75386.1 | |||
| AY158070.1 | AAN76451.1 | |||
| BC111713.1 | AAI11714.1 |
Target Gene Details
Entrez Gene ID: | 5697 |
Gene Name: | peptide YY |
Gene Aliases: |
PYY-I, PYY1 |
Location: |
Chr.17:43952733-44024260 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYY | XM_011525035.1 | XP_011523337.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833454 | Chr.17:43940848 - 44158311 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map