Assay Details
Target Gene Details
Entrez Gene ID: | 8651 |
Gene Name: | suppressor of cytokine signaling 1 |
Gene Aliases: |
CIS1, CISH1, JAB, SOCS-1, SSI-1, SSI1, TIP3 |
Location: |
Chr.16:11254417-11256182 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS1 | NM_003745.1 | 2 | 855 | NP_003736.1 |
| AB000676.1 | 1 | 659 | BAA21537.1 | |
| AB000734.1 | 2 | 855 | BAA23521.1 | |
| AB005043.1 | 1 | 754 | BAA22431.1 | |
| AK127621.1 | 1 | 1423 | ||
| U88326.1 | 1 | 724 | AAB62401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827552 | Chr.16:11254286 - 11257684 on Build GRCh38 | Gain |
|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
| nsv571453 | Chr.16:11218316 - 11302493 on Build GRCh38 | Loss |
|
| nsv525548 | Chr.16:11169999 - 11272732 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map