Assay Details
Target Gene Details
Entrez Gene ID: | 5138 |
Gene Name: | phosphodiesterase 2A |
Gene Aliases: |
CGS-PDE, PDE2A1, PED2A4, cGSPDE |
Location: |
Chr.11:72576140-72674453 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE2A | NM_001143839.3 | 30 | 3428 | NP_001137311.1 |
| NM_001146209.2 | 32 | 3478 | NP_001139681.1 | |
| NM_001243784.1 | 31 | 3490 | NP_001230713.1 | |
| NM_002599.4 | 31 | 3611 | NP_002590.1 | |
| XM_005274040.2 | 31 | 3434 | XP_005274097.1 | |
| XM_011545096.2 | 28 | 3128 | XP_011543398.1 | |
| XM_017017910.1 | 27 | 3844 | XP_016873399.1 | |
| AK092278.1 | ||||
| AK095024.1 | ||||
| AK131525.1 | 21 | 2652 | BAD18664.1 | |
| BC040974.1 | 31 | 3582 | AAH40974.1 | |
| U67733.1 | 31 | 3526 | AAC51320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468633 | Chr.11:72570840 - 72625520 on Build GRCh38 | Loss |
|
| nsv832207 | Chr.11:72561706 - 72758077 on Build GRCh38 | Loss |
|
| nsv509429 | Chr.11:72573921 - 72604515 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map