Assay Details
Target Gene Details
Entrez Gene ID: | 124590 |
Gene Name: | USH1 protein network component sans |
Gene Aliases: |
ANKS4A, SANS |
Location: |
Chr.17:74916083-74923263 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USH1G | NM_001282489.2 | 2 | 1120 | NP_001269418.1 |
| NM_173477.4 | 2 | 1173 | NP_775748.2 | |
| XM_011524296.2 | 2 | 1146 | XP_011522598.1 | |
| AK091243.1 | 2 | 1166 | BAC03619.1 | |
| AK289804.1 | 2 | 1170 | ||
| AK296899.1 | 2 | 1113 | ||
| BC101096.2 | 2 | 1060 | AAI01097.1 | |
| BC101097.1 | 2 | 1060 | AAI01098.1 | |
| BC101098.2 | 2 | 1060 | AAI01099.1 | |
| BC101099.1 | 2 | 1060 | AAI01100.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
| nsv457909 | Chr.17:74889500 - 74982818 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map