Assay Details
Target Gene Details
Entrez Gene ID: | 64651 |
Gene Name: | cysteine and serine rich nuclear protein 1 |
Gene Aliases: |
AXUD1, CSRNP-1, FAM130B, TAIP-3, URAX1 |
Location: |
Chr.3:39141851-39154602 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSRNP1 | NM_001320559.1 | 5 | 2703 | NP_001307488.1 |
| NM_001320560.1 | 5 | 2504 | NP_001307489.1 | |
| NM_033027.3 | 5 | 2572 | NP_149016.2 | |
| XM_017007048.1 | 5 | 5668 | XP_016862537.1 | |
| XM_017007049.1 | 3 | 1956 | XP_016862538.1 | |
| AB053121.1 | 5 | 2549 | BAB61065.1 | |
| AB063301.1 | 5 | 2464 | BAB79450.1 | |
| AL117565.1 | 1 | 775 | CAH10719.1 | |
| BC028213.1 | 3 | 1332 | AAH28213.1 | |
| BC038949.1 | 5 | 2519 | AAH38949.1 | |
| BQ004661.1 | 1 | 630 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590102 | Chr.3:39138418 - 39147352 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map