Assay Details
Target Gene Details
Entrez Gene ID: | 220074 |
Gene Name: | leucine rich transmembrane and O-methyltransferase domain containing |
Gene Aliases: |
CFAP111, DFNB63, LRRC51 |
Location: |
Chr.11:72079982-72110782 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRTOMT | NM_001145307.4 | 4 | 862 | NP_001138779.1 |
| NM_001145308.4 | 2 | 641 | NP_001138780.1 | |
| NM_001145309.3 | 4 | 862 | NP_001138781.1 | |
| NM_001145310.3 | 4 | 862 | NP_001138782.1 | |
| NM_001205138.3 | 3 | 725 | NP_001192067.1 | |
| NM_001271471.2 | 4 | 862 | NP_001258400.1 | |
| NM_001318803.1 | 4 | 819 | NP_001305732.1 | |
| NM_145309.5 | 4 | 862 | NP_660352.1 | |
| NR_026886.3 | 5 | 1042 | ||
| NR_134858.1 | 2 | 641 | ||
| XM_006718473.3 | 4 | 1088 | XP_006718536.1 | |
| XM_006718474.3 | 3 | 1127 | XP_006718537.1 | |
| XM_011544847.2 | 4 | 1316 | XP_011543149.1 | |
| XM_011544848.2 | 5 | 1374 | XP_011543150.1 | |
| XM_017017359.1 | 5 | 1479 | XP_016872848.1 | |
| XM_017017360.1 | 5 | 1331 | XP_016872849.1 | |
| AK298997.1 | 3 | 244 | ||
| AK302772.1 | 2 | 135 | ||
| AK311690.1 | 5 | 512 | ||
| AK313150.1 | 4 | 546 | ||
| AK315892.1 | ||||
| BC012855.1 | 4 | 334 | AAH12855.1 | |
| BI461207.1 | 4 | 305 | ||
| DA004114.1 | 2 | 195 | ||
| EU627066.1 | 4 | 334 | ||
| EU627067.1 | 3 | 284 | ||
| EU627068.1 | 4 | 334 | ||
| EU627069.1 | 4 | 334 | ||
| EU627070.1 | 4 | 334 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv384 | Chr.11:72074236 - 72106174 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map