Assay Details
Target Gene Details
Entrez Gene ID: | 23071 |
Gene Name: | endoplasmic reticulum protein 44 |
Gene Aliases: |
PDIA10, TXNDC4 |
Location: |
Chr.9:99979179-100099052 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP44 | NM_015051.2 | 10 | 1156 | NP_055866.1 |
| AB011145.1 | 10 | 1080 | ||
| AJ344330.2 | 10 | 1067 | CAC87611.1 | |
| AK075024.1 | 10 | 1144 | ||
| AY359048.1 | 10 | 1080 | AAQ89407.1 | |
| BC005374.1 | 10 | 1117 | AAH05374.1 | |
| BX648519.1 | 11 | 1247 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048674 | Chr.9:99989954 - 100275977 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map