Assay Details
Target Gene Details
Entrez Gene ID: | 7389 |
Gene Name: | uroporphyrinogen decarboxylase |
Gene Aliases: |
PCT, UPD |
Location: |
Chr.1:45012133-45015669 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
More Information
Set Membership: |
|

Genomic Map