Assay Details
Target Gene Details
Entrez Gene ID: | 390992 |
Gene Name: | hes family bHLH transcription factor 3 |
Gene Aliases: |
bHLHb43 |
Location: |
Chr.1:6244192-6245578 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HES3 | NM_001024598.3 | NP_001019769.1 | ||
| CN370501.1 | ||||
| CN413592.1 | ||||
| CX755241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8n67 | Chr.1:6220621 - 6264303 on Build GRCh38 | Gain |
|
| nsv831425 | Chr.1:6129291 - 6264418 on Build GRCh38 | Loss |
|
| nsv545236 | Chr.1:6161915 - 6262876 on Build GRCh38 | Loss |
|
| nsv545237 | Chr.1:6240990 - 6250176 on Build GRCh38 | Loss |
|
| nsv508825 | Chr.1:6221275 - 6286549 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map