Assay Details
Target Gene Details
Entrez Gene ID: | 29957 |
Gene Name: | solute carrier family 25 member 24 |
Gene Aliases: |
APC1, SCAMC-1 |
Location: |
Chr.1:108134722-108200358 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A24 | NM_013386.4 | 10 | 2509 | NP_037518.3 |
| NM_213651.2 | 10 | 2420 | NP_998816.1 | |
| XM_017001104.1 | 6 | 1765 | XP_016856593.1 | |
| AF123303.1 | 10 | 2268 | AAF28888.1 | |
| AJ619961.1 | 10 | 2352 | CAF04058.1 | |
| AJ619987.1 | 10 | 2298 | CAF04493.1 | |
| AL050209.1 | 1 | 307 | ||
| BC014519.1 | 10 | 2365 | AAH14519.1 | |
| BC068561.1 | 10 | 2398 | AAH68561.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011093 | Chr.1:107774643 - 108219390 on Build GRCh38 | Gain |
|
| nsv428179 | Chr.1:108076252 - 108554056 on Build GRCh38 | Gain |
|
| dgv248n100 | Chr.1:107385172 - 108286989 on Build GRCh38 | Gain |
|
| dgv469n54 | Chr.1:107776457 - 108233742 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map