Assay Details
Target Gene Details
Entrez Gene ID: | 6183 |
Gene Name: | mitochondrial ribosomal protein S12 |
Gene Aliases: |
MPR-S12, MT-RPS12, RPMS12, RPS12, RPSM12 |
Location: |
Chr.19:38930708-38933020 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS12 | NM_021107.1 | 2 | 659 | NP_066930.1 |
| NM_033362.3 | 3 | 628 | NP_203526.1 | |
| NM_033363.1 | 3 | 555 | NP_203527.1 | |
| BC001617.1 | 3 | 510 | AAH01617.1 | |
| BC006424.1 | 3 | 788 | AAH06424.1 | |
| BC007499.2 | 3 | 381 | AAH07499.1 | |
| CR407609.1 | 2 | 318 | CAG28537.1 | |
| Y11681.1 | 2 | 659 | CAA72377.1 |
More Information
Set Membership: |
|

Genomic Map