Assay Details
Target Gene Details
Entrez Gene ID: | 9113 |
Gene Name: | large tumor suppressor kinase 1 |
Gene Aliases: |
WARTS, wts |
Location: |
Chr.6:149658153-149718256 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LATS1 | NM_001270519.1 | 1 | 175 | NP_001257448.1 |
| NM_004690.3 | 1 | 175 | NP_004681.1 | |
| XM_006715603.3 | XP_006715666.1 | |||
| XM_011536252.2 | XP_011534554.1 | |||
| XM_017011474.1 | XP_016866963.1 | |||
| XM_017011476.1 | XP_016866965.1 | |||
| XM_017011477.1 | XP_016866966.1 | |||
| XM_017011478.1 | XP_016866967.1 | |||
| XM_017011479.1 | XP_016866968.1 | |||
| XM_017011481.1 | XP_016866970.1 | |||
| XM_017011482.1 | XP_016866971.1 | |||
| AF104413.1 | 1 | 175 | AAD16882.1 | |
| BC002767.1 | AAH02767.1 | |||
| BC015665.2 | AAH15665.1 |
Target Gene Details
Entrez Gene ID: | 645967 |
Gene Name: | uncharacterized LOC645967 |
Gene Aliases: |
- |
Location: |
Chr.6:149717621-149718896 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC645967 | NR_134652.1 | 1 | 462 | |
| AB593152.1 | 1 | 462 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2732861 | Chr.6:149681836 - 149764933 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs542474056] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map