Assay Details
Target Gene Details
Entrez Gene ID: | 29965 |
Gene Name: | cell death-inducing p53 target 1 |
Gene Aliases: |
C16orf5, CDIP, I1, LITAFL |
Location: |
Chr.16:4510675-4538815 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDIP1 | NM_001199054.1 | 6 | 2375 | NP_001185983.1 |
| NM_001199055.1 | 6 | 2226 | NP_001185984.1 | |
| NM_001199056.1 | 6 | 2106 | NP_001185985.1 | |
| NM_013399.2 | 6 | 2343 | NP_037531.2 | |
| AF131218.1 | 5 | 2422 | AAF26619.1 | |
| AL833853.1 | 6 | 2375 | CAD38712.1 | |
| BC007604.1 | 2 | 1877 | AAH07604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map