Assay Details
Target Gene Details
Entrez Gene ID: | 6461 |
Gene Name: | SH2 domain containing adaptor protein B |
Gene Aliases: |
bA3J10.2 |
Location: |
Chr.9:37915898-38069213 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHB | NM_003028.2 | 6 | 4009 | NP_003019.2 |
| AK055947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469814 | Chr.9:37777286 - 37950154 on Build GRCh38 | Gain |
|
| esv2738434 | Chr.9:37917944 - 37918684 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map