Assay Details
Target Gene Details
Entrez Gene ID: | 152641 |
Gene Name: | WWC2 antisense RNA 2 |
Gene Aliases: |
C4orf38 |
Location: |
Chr.4:183097021-183099199 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC2-AS2 | NR_024008.1 | 1 | 1420 | |
| AK054839.1 | 1 | 1420 | BAB70812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822872 | Chr.4:183097250 - 183100919 on Build GRCh38 | Gain |
|
| nsv1016376 | Chr.4:182859067 - 183262599 on Build GRCh38 | Gain |
|
| dgv9415n54 | Chr.4:183095835 - 183099302 on Build GRCh38 | Loss |
|

Genomic Map