Assay Details
Target Gene Details
Entrez Gene ID: | 79726 |
Gene Name: | WD repeat domain 59 |
Gene Aliases: |
FP977 |
Location: |
Chr.16:74873568-74985146 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDR59 | NM_030581.3 | 26 | 3400 | NP_085058.3 |
| XM_005256146.3 | 27 | 3461 | XP_005256203.1 | |
| XM_011523331.2 | 25 | 3221 | XP_011521633.1 | |
| XM_011523332.2 | 21 | 2909 | XP_011521634.1 | |
| XM_017023667.1 | 25 | 3261 | XP_016879156.1 | |
| XM_017023668.1 | 24 | 3037 | XP_016879157.1 | |
| XM_017023669.1 | 25 | 3109 | XP_016879158.1 | |
| XM_017023670.1 | 24 | 3142 | XP_016879159.1 | |
| AB067510.1 | 21 | 2873 | BAB67816.1 | |
| AF370390.1 | 14 | 2332 | AAQ15226.1 | |
| AK022332.1 | 9 | 1832 | BAB14015.1 | |
| AK091316.1 | 14 | 2180 | ||
| AL832078.1 | 11 | 1808 | ||
| BC014887.2 | 20 | 2792 | AAH14887.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833280 | Chr.16:74821913 - 75043329 on Build GRCh38 | Loss |
|
| dgv3019n100 | Chr.16:74834429 - 74955590 on Build GRCh38 | Loss |
|
| nsv572978 | Chr.16:74839676 - 74956106 on Build GRCh38 | Loss |
|
| nsv526213 | Chr.16:74839615 - 74956163 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map