Assay Details
Target Gene Details
Entrez Gene ID: | 84309 |
Gene Name: | nudix hydrolase 16 like 1 |
Gene Aliases: |
SDOS |
Location: |
Chr.16:4691340-4695859 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT16L1 | NM_001193452.1 | 3 | 984 | NP_001180381.1 |
| NM_032349.3 | 3 | 914 | NP_115725.1 | |
| XM_005255633.4 | 2 | 3986 | XP_005255690.1 | |
| AK092642.1 | 2 | 1608 | BAC03933.1 | |
| BC006223.2 | 3 | 892 | AAH06223.1 | |
| BU739233.1 | 1 | 454 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160333 | Chr.16:4570296 - 4709737 on Build GRCh38 | Deletion |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv1050237 | Chr.16:4641028 - 4744872 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map