Assay Details
Target Gene Details
Entrez Gene ID: | 1147 |
Gene Name: | conserved helix-loop-helix ubiquitous kinase |
Gene Aliases: |
IKBKA, IKK-alpha, IKK1, IKKA, NFKBIKA, TCF16 |
Location: |
Chr.10:100188298-100229610 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHUK | NM_001278.4 | NP_001269.3 | ||
| NM_001320928.1 | NP_001307857.1 | |||
| XM_017015611.1 | XP_016871100.1 | |||
| XM_017015612.1 | XP_016871101.1 | |||
| XM_017015613.1 | XP_016871102.1 | |||
| AB209028.1 | BAD92265.1 | |||
| AF009225.1 | AAC51671.1 | |||
| AF012890.1 | AAC51662.1 | |||
| AF080157.1 | AAD08996.1 | |||
| BC092514.1 | AAH92514.1 | |||
| U22512.1 | AAC50713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516860 | Chr.10:100198475 - 100246924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map