Assay Details
Target Gene Details
Entrez Gene ID: | 27178 |
Gene Name: | interleukin 37 |
Gene Aliases: |
FIL1, FIL1(ZETA), FIL1Z, IL-1F7, IL-1H, IL-1H4, IL-1RP1, IL-37, IL1F7, IL1H4, IL1RP1 |
Location: |
Chr.2:112908889-112918882 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL37 | NM_014439.3 | NP_055254.2 | ||
| NM_173202.1 | NP_775294.1 | |||
| NM_173203.1 | NP_775295.1 | |||
| NM_173204.1 | NP_775296.1 | |||
| NM_173205.1 | NP_775297.1 | |||
| XM_011510963.2 | XP_011509265.1 | |||
| XM_011510964.2 | XP_011509266.1 | |||
| AF167368.1 | AAG29344.1 | |||
| AF200496.1 | AAF69252.1 | |||
| AF251118.1 | AAG14420.1 | |||
| BC020637.1 | AAH20637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003372 | Chr.2:112913313 - 112925044 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map