Assay Details
Target Gene Details
Entrez Gene ID: | 2017 |
Gene Name: | cortactin |
Gene Aliases: |
EMS1 |
Location: |
Chr.11:70398506-70436584 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTTN | NM_001184740.1 | NP_001171669.1 | ||
| NM_005231.3 | NP_005222.2 | |||
| NM_138565.2 | NP_612632.1 | |||
| XM_006718447.2 | XP_006718510.1 | |||
| XM_006718448.3 | XP_006718511.1 | |||
| XM_017017312.1 | XP_016872801.1 | |||
| AK222613.1 | BAD96333.1 | |||
| AK291097.1 | ||||
| AU124856.1 | ||||
| BC008799.2 | AAH08799.1 | |||
| BC033889.1 | AAH33889.1 | |||
| DA716772.1 | ||||
| M98343.1 | AAA58455.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73516264] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map