Assay Details
Target Gene Details
Entrez Gene ID: | 5708 |
Gene Name: | proteasome 26S subunit, non-ATPase 2 |
Gene Aliases: |
P97, RPN1, S2, TRAP2 |
Location: |
Chr.3:184299098-184309054 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD2 | NM_001278708.1 | NP_001265637.1 | ||
| NM_001278709.1 | NP_001265638.1 | |||
| NM_002808.4 | NP_002799.3 | |||
| AB209843.1 | BAD93080.1 | |||
| AK095245.1 | ||||
| AK297256.1 | ||||
| AK301759.1 | ||||
| AK302177.1 | ||||
| AK315474.1 | ||||
| AL162014.1 | CAB82366.2 | |||
| BC002368.2 | AAH02368.1 | |||
| BC002997.1 | AAH02997.1 | |||
| BC007897.2 | AAH07897.1 | |||
| BT009736.1 | AAP88738.1 | |||
| D78151.1 | BAA11226.1 | |||
| U12596.1 | AAA87705.1 | |||
| U18247.1 | AAA57339.1 | |||
| X86446.1 | CAA60167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422400 | Chr.3:184299495 - 184425713 on Build GRCh38 | Duplication |
|
| nsv829813 | Chr.3:184264997 - 184427996 on Build GRCh38 | Loss |
|
| dgv961e214 | Chr.3:184286928 - 184306834 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map