Assay Details
Target Gene Details
Entrez Gene ID: | 4700 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A6 |
Gene Aliases: |
B14, CI-B14, LYRM6, NADHB14 |
Location: |
Chr.22:42085526-42090884 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA6 | NM_002490.3 | 3 | 995 | NP_002481.2 |
| BC002772.1 | 3 | 856 | AAH02772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508737 | Chr.22:42052007 - 42134336 on Build GRCh38 | Insertion |
|
| nsv834210 | Chr.22:42057329 - 42209100 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map