Assay Details
Target Gene Details
Entrez Gene ID: | 59343 |
Gene Name: | SUMO1/sentrin/SMT3 specific peptidase 2 |
Gene Aliases: |
AXAM2, SMT3IP2 |
Location: |
Chr.3:185586228-185631101 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP2 | NM_021627.2 | 3 | 411 | NP_067640.2 |
| XM_005247690.2 | 3 | 426 | XP_005247747.2 | |
| XM_005247691.2 | 3 | 357 | XP_005247748.1 | |
| XM_017006973.1 | 3 | 357 | XP_016862462.1 | |
| AF151697.2 | 3 | 414 | AAG15309.2 | |
| AK027599.1 | 3 | 352 | BAB55222.1 | |
| AK296744.1 | 3 | 343 | ||
| AK296772.1 | 3 | 359 | ||
| AK298628.1 | 3 | 238 | ||
| AK304402.1 | 3 | 480 | ||
| AL834380.1 | 3 | 235 | CAD39043.1 | |
| BC040609.1 | 3 | 297 | AAH40609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4148 | Chr.3:185592906 - 185625116 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map