Assay Details
Target Gene Details
Entrez Gene ID: | 147184 |
Gene Name: | transmembrane protein 99 |
Gene Aliases: |
- |
Location: |
Chr.17:40819092-40836274 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM99 | NM_001195386.1 | 3 | 788 | NP_001182315.1 |
| NM_001195387.1 | 3 | 772 | NP_001182316.1 | |
| NM_145274.3 | 3 | 780 | NP_660317.2 | |
| XM_005257089.4 | 4 | 977 | XP_005257146.1 | |
| XM_017024253.1 | 4 | 945 | XP_016879742.1 | |
| AI301318.1 | 1 | 101 | ||
| AK097454.1 | 3 | 721 | BAC05059.1 | |
| AK298616.1 | 3 | 723 | ||
| BC015365.1 | 3 | 759 | AAH15365.1 | |
| BG530583.1 | 3 | 758 |

Genomic Map