Assay Details
Target Gene Details
Entrez Gene ID: | 54814 |
Gene Name: | glutaminyl-peptide cyclotransferase like |
Gene Aliases: |
gQC |
Location: |
Chr.19:45692402-45703990 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QPCTL | NM_001163377.1 | 6 | 1177 | NP_001156849.1 |
| NM_017659.3 | 7 | 1459 | NP_060129.2 | |
| AK000091.1 | 7 | 1268 | BAA90938.1 | |
| AK172764.1 | 7 | 1267 | BAD18747.1 | |
| AK222636.1 | 7 | 1252 | BAD96356.1 | |
| BC011553.1 | 6 | 963 | AAH11553.1 | |
| BU927708.1 | 1 | 84 |
More Information
Set Membership: |
|

Genomic Map