Assay Details
Target Gene Details
Entrez Gene ID: | 64689 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
GOLPH5, GRASP65, P65 |
Location: |
Chr.3:39096599-39108363 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GORASP1 | NM_001278789.1 | NP_001265718.1 | ||
| NM_001278790.1 | NP_001265719.1 | |||
| NM_031899.3 | NP_114105.1 | |||
| NR_103866.1 | ||||
| NR_103867.1 | ||||
| XM_006713301.3 | XP_006713364.1 | |||
| XM_011534020.2 | XP_011532322.1 | |||
| XM_011534021.2 | XP_011532323.1 | |||
| XM_017007050.1 | XP_016862539.1 | |||
| XM_017007051.1 | XP_016862540.1 | |||
| XM_017007052.1 | XP_016862541.1 | |||
| AJ409349.1 | CAC35160.1 | |||
| AK001574.1 | ||||
| AK057043.1 | ||||
| AK091168.1 | BAC03598.1 | |||
| AK124755.1 | ||||
| AK300271.1 | ||||
| AK303847.1 | ||||
| BC008928.2 | AAH08928.1 | |||
| BC075854.1 | AAH75854.1 | |||
| BC103826.1 | AAI03827.1 | |||
| BC103827.1 | AAI03828.1 | |||
| DB031221.1 |
More Information
Set Membership: |
|

Genomic Map