Assay Details
Target Gene Details
Entrez Gene ID: | 9962 |
Gene Name: | solute carrier family 23 member 2 |
Gene Aliases: |
NBTL1, SLC23A1, SVCT2, YSPL2, hSVCT2 |
Location: |
Chr.20:4852356-5010293 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC23A2 | NM_005116.5 | 17 | 6164 | NP_005107.4 |
| NM_203327.1 | 17 | 6182 | NP_976072.1 | |
| XM_011529414.2 | 17 | 6169 | XP_011527716.1 | |
| XM_011529415.2 | 16 | 5990 | XP_011527717.1 | |
| XM_011529416.2 | 16 | 6299 | XP_011527718.1 | |
| XM_011529417.2 | 16 | 6046 | XP_011527719.1 | |
| XM_017028171.1 | 17 | 6203 | XP_016883660.1 | |
| XM_017028172.1 | 17 | 6207 | XP_016883661.1 | |
| XM_017028173.1 | 17 | 6187 | XP_016883662.1 | |
| XM_017028174.1 | 17 | 6082 | XP_016883663.1 | |
| XM_017028175.1 | 13 | 5518 | XP_016883664.1 | |
| XM_017028176.1 | 14 | 5836 | XP_016883665.1 | |
| XM_017028177.1 | 14 | 5823 | XP_016883666.1 | |
| XM_017028178.1 | 16 | 6089 | XP_016883667.1 | |
| XM_017028179.1 | 16 | 6076 | XP_016883668.1 | |
| XM_017028180.1 | 12 | 5400 | XP_016883669.1 | |
| AB075494.1 | 1 | 1035 | ||
| AK025668.1 | ||||
| D87075.2 | 17 | 6164 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645074 | Chr.20:4205961 - 5037636 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map