Assay Details
Target Gene Details
Entrez Gene ID: | 2232 |
Gene Name: | ferredoxin reductase |
Gene Aliases: |
ADXR |
Location: |
Chr.17:74862497-74873031 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FDXR | NM_001258012.3 | 3 | 438 | NP_001244941.2 |
| NM_001258013.3 | 4 | 402 | NP_001244942.2 | |
| NM_001258014.3 | 3 | 309 | NP_001244943.2 | |
| NM_001258015.2 | 3 | 312 | NP_001244944.1 | |
| NM_001258016.3 | 2 | 254 | NP_001244945.2 | |
| NM_004110.5 | 3 | 309 | NP_004101.3 | |
| NM_024417.4 | 3 | 309 | NP_077728.3 | |
| NR_047576.2 | 3 | 409 | ||
| XM_006721772.1 | 3 | 257 | XP_006721835.1 | |
| XM_011524527.1 | 3 | 257 | XP_011522829.1 | |
| XM_011524528.1 | 3 | 346 | XP_011522830.1 | |
| XM_011524529.1 | 3 | 349 | XP_011522831.1 | |
| XM_011524531.2 | 2 | 242 | XP_011522833.1 | |
| XM_011524532.2 | 2 | 249 | XP_011522834.1 | |
| XM_011524533.2 | 2 | 235 | XP_011522835.1 | |
| AK094120.1 | 4 | 507 | ||
| AK098648.1 | 3 | 246 | ||
| AK223530.1 | 3 | 239 | BAD97250.1 | |
| AK293208.1 | 4 | 347 | ||
| AK293210.1 | 3 | 346 | ||
| AK295307.1 | 3 | 438 | ||
| AK298908.1 | 3 | 255 | ||
| AK301779.1 | 3 | 246 | ||
| AK301977.1 | 2 | 254 | ||
| BC002960.1 | 3 | 248 | AAH02960.1 | |
| BC063493.1 | 3 | 247 | AAH63493.1 | |
| BT006684.1 | 3 | 222 | AAP35330.1 | |
| DA267443.1 | 4 | 402 | ||
| J03826.1 | 3 | 242 | AAB59497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map