Assay Details
Target Gene Details
Entrez Gene ID: | 65123 |
Gene Name: | integrator complex subunit 3 |
Gene Aliases: |
C1orf193, C1orf60, INT3, SOSS-A, SOSSA |
Location: |
Chr.1:153728050-153774808 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS3 | NM_001324475.1 | NP_001311404.1 | ||
| NM_023015.4 | NP_075391.3 | |||
| XM_005245459.2 | XP_005245516.1 | |||
| XM_006711490.3 | XP_006711553.1 | |||
| XM_011509906.2 | XP_011508208.1 | |||
| AK290025.1 | ||||
| AL832133.1 | CAH10398.1 | |||
| BC105092.1 | AAI05093.1 | |||
| BC105094.1 | AAI05095.1 | |||
| BC116458.1 | ||||
| BX640786.1 | CAE45876.1 | |||
| BX640950.1 | CAE45974.1 | |||
| CR627233.1 | CAH10366.1 | |||
| CR749212.1 | CAH18069.1 | |||
| CR749376.1 | CAH18229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464028 | Chr.1:153649744 - 153750561 on Build GRCh38 | Gain |
|
| dgv582n54 | Chr.1:153639657 - 153750561 on Build GRCh38 | Gain |
|
| nsv547941 | Chr.1:153736430 - 153828622 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map